missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SCGB3A2 Protein
A cDNA sequence encoding the SCGB3A2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13429-2ug
Additional Details : Weight : 0.01000kg
Specifications
SCGB3A2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MFLINKVPLP VDKLAPLPLD NILPFMDPLK LLLKTLGISV EHLVEGLRKC VNELGPEASE AVKKLLEALS HLVGGSGGSF LINKVPLPVD KLAPLPLDNI LPFMDPLKLL LKTLGISVEH LVEGLRKCVN ELGPEASEAV KKLLEALSHL VHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SCGB3A2 solution at a concentration of 0.4 mg/ml in phosphate buffered saline (PBS) pH 8.0. |