missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Selenoprotein 1
A DNA sequence encoding the Homo sapiens (Human) Selenoprotein P, was expressed in the hosts and tags indicated.
203.00 € - 2285.00 €
Specifications
Accession Number | NP_001078955.1 |
---|---|
Gene ID (Entrez) | 6414 |
Name | Selenoprotein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16034920
|
enQuireBio™
QP6665-EC-10UG |
10 μg |
203.00 €
10µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024920
|
enQuireBio™
QP6665-EC-100UG |
100 μg |
505.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16044920
|
enQuireBio™
QP6665-EC-1MG |
1 mg |
2285.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16054920
|
enQuireBio™
QP6665-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16064920
|
enQuireBio™
QP6665-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16074920
|
enQuireBio™
QP6665-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001078955.1 | |
Selenoprotein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6414 | |
Research Use Only | |
SEPP1 | |
Recombinant Protein | |
E. coli | |
ESQDQSSLCK QPPAWSIRDQ DPMLNSNGSV TVVALLQASS YLCILQASKL EDLRVKLKKE GYSNISYIVV NHQGISSRLK YTHLKNKVSE HIPVYQQEEN QTDVWTLLNG SKDDFLIYDR CGRLVYHLGL PFSFLTFPYV EEAIKIAYCE KKCGNCSLTT LKDEDFCKRV SLATVDKTVE TPSPHYHHEH HHNHGHQHLG SSELSENQQP GAPNAPTHPA PPGLHHHHKH KGQHRQGHPE NRDMPASEDL QDLQKKLCRK RCINQLLCKL PTDSELAPRS SCCHCRHLIF EKTGSAITSQ CKENLPSLCS SQGLRAEENI TESCQSRLPP AASQISQQLI PTEASASSRS KNQAKKSESP SN | |
Greater than 90% as determined by SDS-PAGE. |