missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SGTA Protein

Product Code. 15973719
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
This item is not returnable. View return policy

Product Code. 15973719

Brand: enQuireBio™ QP134845ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the SGTA was constructed and used to recombinantly synthesize the protein.

Specifications

Name SGTA Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MDNKKRLAYA IIQFLHDQLR HGGLSSDAQE SLEVAIQCLE TAFGVTVEDS DLALPQTLPE IFEAAATGKE MPQDLRSPAR TPPSEEDSAE AERLKTEGNE QMKVENFEAA VHFYGKAIEL NPANAVYFCN RAAAYSKLGN YAGAVQDCER AICIDPAYSK AYGRMGLALS SLNKHVEAVA YYKKALELDP DNETYKSNLK IAELKLREAP SPTGGVGSFD IAGLLNNPGF MSMASNLMNN PQIQQLMSGM ISGGNNPLGT PGTSPSQNDL ASLIQAGQQF AQQMQQQNPE LIEQLRSQIR SRTPSASNDD QQELEHHHHH H
Buffer SGTA Human solution containing 20mM Trsi HCL pH 8.0, 1mM DTT and 10% glycerol.
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.