missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SH3GL3 Protein
A cDNA sequence encoding the SH3GL3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13491-1mg
Additional Details : Weight : 0.01000kg
Specifications
SH3GL3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSVAGLK KQFHKASQLF SEKISGAEGT KLDDEFLDME RKIDVTNKVV AEILSKTTEY LQPNPAYRAK LGMLNTVSKI RGQVKTTGYP QTEGLLGDCM LKYGKELGED STFGNALIEV GESMKLMAEV KDSLDINVKQ TFIDPLQLLQ DKDLKEIGHH LKKLEGRRLD YDYKKKRVGK IPDEEVRQAV EKFEESKELA ERSMFNFLEN DVEQVSQLAV FIEAALDYHR QSTEILQELQ SKLQMRISAA SSVPRREYKP RPVKRSSSEL NGVSTTSVVK TTGSNIPMDQ PCCRGLYDFE PENQGELGFK EGDIITLTNQ IDENWYEGMI HGESGFFPIN YVEVIVPLPQ | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SH3GL3 protein solution (1 mg/ml) containing PBS (pH 7.4), 10% glycerol and 1mM DTT. |