missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SHH / Sonic hedgehog Protein
Recombinant Protein
Brand: enQuireBio™ QP5292-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000184.1 | |
SHH / Sonic hedgehog Protein | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 5 ug/mL in the CCL-226 cell proliferation assay. The bioactivity acceptance criteria is set at 200 units/mg. | |
Human | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
6469 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
SHH | |
Recombinant Protein | |
E. coli | |
MIIGPGRGFG KRRHPKKLTP LAYKQFIPNV AEKTLGASGR YEGKISRNSE RFKELTPNYN PDIIFKDEEN TGADRLMTQR CKDKLNALAI SVMNQWPGVK LRVTEGWDED GHHSEESLHY EGRALDITTS DRDRSKYGML ARLAVEAGFD WVYYESKAHI HCSVKAENSV AAKSGG |