missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Sialic acid-binding Ig-like lectin 15 Protein

A DNA sequence encoding the Homo sapiens (Human) Sialic acid-binding Ig-like lectin 15, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7774-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16050711

  • 2285.00 € / 1mg
Estimated Shipment: 04-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_998767.1
Sialic acid-binding Ig-like lectin 15 Protein
Research Use Only
SIGLEC15
Recombinant Protein
E. coli
FVRTKIDTTE NLLNTEVHSS PAQRWSMQVP PEVSAEAGDA AVLPCTFTHP HRHYDGPLTA IWRAGEPYAG PQVFRCAAAR GSELCQTALS LHGRFRLLGN PRRNDLSLRV ERLALADDRR YFCRVEFAGD VHDRYESRHG VRLHVTAAPR IVNISVLPSP AHAFRALCTA EGEPPPALAW SGPALGNSLA AVRSPREGHG HLVTAELPAL THDGRYTCTA ANSLGRSEAS VYLFRFHGAS GAST
Greater than 90% as determined by SDS-PAGE.
284266
1 mg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers