missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SIRP alpha / CD172a Protein

A DNA sequence encoding the Homo sapiens (Human) SIRP alpha / CD172a, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6684-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16065080

  • 2285.00 € / 1mg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001035111.1
SIRP alpha / CD172a Protein
Research Use Only
SIRPA
Recombinant Protein
E. coli
EEELQVIQPD KSVLVAAGET ATLRCTATSL IPVGPIQWFR GAGPGRELIY NQKEGHFPRV TTVSDLTKRN NMDFSIRIGN ITPADAGTYY CVKFRKGSPD DVEFKSGAGT ELSVRAKPSA PVVSGPAARA TPQHTVSFTC ESHGFSPRDI TLKWFKNGNE LSDFQTNVDP VGESVSYSIH STAKVVLTRE DVHSQVICEV AHVTLQGDPL RGTANLSETI RVPPTLEVTQ QPVRAENQVN VTCQVRKFYP QRLQLTWLEN GNVSRTETAS TVTENKDGTY NWMSWLLVNV SAHRDDVKLT CQVEHDGQPA VSKSHDLKVS AHPKEQGSNT AAENTGSNER NIY
Greater than 90% as determined by SDS-PAGE.
140885
1 mg
Not determined.
Extracellular Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers