missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SLAMF1 Protein
A cDNA sequence encoding the SLAMF1 was constructed and used to recombinantly synthesize the protein.
Marke: enQuireBio™ QP13518-HIS-EC-2ug
Additional Details : Weight : 0.01000kg
Spezifikation
6504 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSASYGTGG RMMNCPKILR QLGSKVLLPL TYERINKSMN KSIHIVVTMA KSLENSVENK IVSLDPSEAG PPRYLGDRYK FYLENLTLGI RESRKEDEGW YLMTLEKNVS VQRFCLQLRL YEQVSTPEIK VLNKTQENGT CTLILGCTVE KGDHVAYSWS EKAGTHPLNP ANSSHLLSLT LGPQHADNIY ICTVSNPISN NSQTFSPWPG CRTDPSETKP | |
Greater than 85.0% as determined by SDS-PAGE. |
SLAMF1 Protein | |
Research Use Only | |
SLAMF1 | |
Human | |
His | |
SLAMF1 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M UREA and 10% glycerol. |