missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Small Nuclear Ribonucleoprotein, 70kDa Protein

Product Code. 15985459
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15985459

Brand: enQuireBio™ QP140315ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Small Nuclear Ribonucleoprotein, 70kDa was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 6625
Name Small Nuclear Ribonucleoprotein, 70kDa Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol SNRNP70
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MTQFLPPNLL ALFAPRDPIP YLPPLEKLPH EKHHNQPYCG IAPYIREFED PRDAPPPTRA ETREERMERK RREKIERRQQ EVETELKMWD PHNDPNAQGD AFKTLFVARV NYDTTESKLR REFEVYGPIK RIHMVYSKRS GKPRGYAFIE YEHERDMHSA YKHADGKKID GRRVLVDVER GRTVKGWRPR RLGGGLGGTR RGGADVNIRH SGRDDTSRYD ERPGPSPLPH RDRDRDRERE RRERSRERDK ERERRRSRSR DRRRRSRSRD KEERRRSRER SKDKDRDRKR RSSRSRERAR RERERKEELR GGGGDMAEPS EAGDAPPDDG PPGELGPDGP DGPEEKGRDR DRERRRSHRS ERERRRDRDR DRDRDREHKR GERGSERGRD EARGGGGGQD NGLEGLGNDS RDMYMESEGG DGYLAPENGY LMEAAPE
Buffer U1-snRNP is supplied in 20mM HEPES buffer pH 7.5, 0.01mM EDTA and 0.02% SDS.
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.