missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNRNP25 Protein
A cDNA sequence encoding the SNRNP25 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13540-20ug
Additional Details : Weight : 0.01000kg
Specifications
SNRNP25 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MDVFQEGLAM VVQDPLLCDL PIQVTLEEVN SQIALEYGQA MTVRVCKMDG EVMPVVVVQS ATVLDLKKAI QRYVQLKQER EGGIQHISWS YVWRTYHLTS AGEKLTEDRK KLRDYGIRNR DEVSFIKKLR QK | |
Greater than 85.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
SNRNP25 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 2mM DTT and 0.2M NaCl. |