missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SNURF Protein
A cDNA sequence encoding the SNURF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13555-1mg
Spécification
SNURF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMERARDR LHLRRTTEQH VPEVEVQVKR RRTASLSNQE CQLYPRRSQQ QQVPVVDFQA ELRQAFLAET PRGG | |
Greater than 90% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The SNURF solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 1mM DTT and 40% glycerol. |