missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SPRY4 Protein
Click to view available options
Quantity:
1 mg
25 μg
5 μg
Unit Size:
1mg
25µg
5µg
Specifica
Specifica
| Name | SPRY4 Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMEPPIPQ SAPLTPNSVM VQPLLDSRMS HSRLQHPLTI LPIDQVKTSH VENDYIDNPS LALTTGPKRT RGGAPELAPT PARCDQDVTH HWISFSGRPS SVSSSSSTSS DQRLLDHMAP PPVADQASPR AVRIQPKVVH CQPLDLKGPA VPPELDKHFL LCEACGKCKC KECASPRTLP SCWVCNQECL CSAQTLVNYG TCMCLVQGIF YHCTNEDDEG SCADHPCSCS RSNCCARWSF MGALSVVLPC LLCYLPATGC VKLAQRGYDR LRRPGCRCKH TNSVICKAAS GDAKTSRPDK PF |
| Buffer | The SPRY4 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |
| Vedi altri risultati |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto