missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SRA1 Protein
A cDNA sequence encoding the SRA1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13589-100ug
Additional Details : Weight : 0.01000kg
Specifications
10011 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSVGSGPAS GVEPTSFPVE SEAVMEDVLR PLEQALEDCR GHTRKQVCDD ISRRLALLQE QWAGGKLSIP VKKRMALLVQ ELSSHRWDAA DDIHRSLMVD HVTEVSQWMV GVKRLIAEKR SLFSEEAANE EKSAATAEKN HTIPGFQQAS | |
Greater than 85.0% as determined by SDS-PAGE. |
SRA1 Protein | |
Research Use Only | |
SRA1 | |
Human | |
His | |
SRA1 protein solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0) , 0.1MNaCl ,10% glycerol and 1mM DTT. |