missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STAC Protein

A cDNA sequence encoding the STAC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13605-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954119

  • 203.00 € / 20µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

STAC Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMIPPSSP REDGVDGLPK EAVGAEQPPS PASTSSQESK LQKLKRSLSF KTKSLRSKSA DNFFQRTNSE DMKLQAHMVA EISPSSSPLP APGSLTSTPA RAGLHPGGKA HAFQEYIFKK PTFCDVCNHM IVGTNAKHGL RCKACKMSIH HKCTDGLAPQ RCMGKLPKGF RRYYSSPLLI HEQFGCIKEV MPIACGNKVD PVYETLRFGT SLAQRTKKGS SGSGSDSPHR TSTSDLVEVP EEANGPGGGY DLRKRSNSVF TYPENGTDDF RDPAKNINHQ GSLSKDPLQM NTYVALYKFV PQENEDLEMR PGDIITLLED SNEDWWKGKI QDRIGFFPAN FVQRLQQNEK IFRCVRTFIG CKEQGQITLK ENQICVSSEE EQDGFIRVLS GKKKGLIPLD VLENI
Greater than 85.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
STAC protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers