missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human STAP1 Protein

A cDNA sequence encoding the STAP1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13608-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934129

  • 3915.00 € / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

STAP1 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMMAKKP PKPAPRRIFQ ERLKITALPL YFEGFLLIKR SGYREYEHYW TELRGTTLFF YTDKKSIIYV DKLDIVDLTC LTEQNSTEKN CAKFTLVLPK EEVQLKTENT ESGEEWRGFI LTVTELSVPQ NVSLLPGQVI KLHEVLEREK KRRIETEQST SVEKEKEPTE DYVDVLNPMP ACFYTVSRKE ATEMLQKNPS LGNMILRPGS DSRNYSITIR QEIDIPRIKH YKVMSVGQNY TIELEKPVTL PNLFSVIDYF VKETRGNLRP FICSTDENTG QEPSMEGRSE KLKKNPHIA
Greater than 95.0% as determined by SDS-PAGE.
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
STAP1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.1M NaCl and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers