missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STK17B Protein
A cDNA sequence encoding the STK17B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13618-100ug
Additional Details : Weight : 0.01000kg
Specifications
9262 | |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMSRRRFD CRSISGLLTT TPQIPIKMEN FNNFYILTSK ELGRGKFAVV RQCISKSTGQ EYAAKFLKKR RRGQDCRAEI LHEIAVLELA KSCPRVINLH EVYENTSEII LILEYAAGGE IFSLCLPELA EMVSENDVIR LIKQILEGVY YLHQNNIVHLDLKPQNILLS SIYPLGDIKI VDFGMSRKIG HACELREIMG TPEYLAPEIL NYDPITTATD MWNIGIIAYM LLTHTSPFVG EDNQETYLNI SQVNVDYSEE TFSSVSQLAT DFIQSLLVKN PEKRPTAEIC LSHSWLQQWD FENLFHPEET SSSSQTQDHS VRSSEDKTSK SSCNGTCGDREDKENIPEDS SMVSKRFRFD DSLPNPHELV SDLLC | |
Greater than 90.0% as determined by SDS-PAGE. |
STK17B Protein | |
Research Use Only | |
STK17B | |
Human | |
His | |
STK17B protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 2mM DTT. |