missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human STOM / EPB72 Protein
A DNA sequence encoding the Homo sapiens (Human) STOM / EPB72, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6740-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_004090.4 | |
STOM / EPB72 Protein | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
2040 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
STOM | |
Recombinant Protein | |
E. coli | |
MAEKRHTRDS EAQRLPDSFK DSPSKGLGPC GWILVAFSFL FTVITFPISI WMCIKIIKEY ERAIIFRLGR ILQGGAKGPG LFFILPCTDS FIKVDMRTIS FDIPPQEILT KDSVTISVDG VVYYRVQNAT LAVANITNAD SATRLLAQTT LRNVLGTKNL SQILSDREEI AHNMQSTLDD ATDAWGIKVE RVEIKDVKLP VQLQRAMAAE AEASREARAK VIAAEGEMNA SRALKEASMV ITESPAALQL RYLQTLTTIA AEKNSTIVFP LPIDMLQGII GAKHSHLG |