missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SUMO1 Protein

A cDNA sequence encoding the SUMO1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13648-HIS-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15984259

  • 203.00 € / 20µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

SUMO1 Protein
Research Use Only
Recombinant Protein
E. coli
MSDQEAKPST EDLGDKKEGE YIKLKVIGQD SSEIHFKVKM TTHLKKLKES YCQRQGVPMN SLRFLFEGQR IADNHTPKEL GMEEEDVIEV YQEQTGGHST V
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
10mM sodium chloride, 100mM imidazole, 0.5mM PMSF, 1mM DTT and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers