missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SUMO1 Protein
A cDNA sequence encoding the SUMO1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13648-HIS-5ug
Additional Details : Weight : 0.01000kg
Specifications
SUMO1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSDQEAKPST EDLGDKKEGE YIKLKVIGQD SSEIHFKVKM TTHLKKLKES YCQRQGVPMN SLRFLFEGQR IADNHTPKEL GMEEEDVIEV YQEQTGGHST V | |
Greater than 98.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
10mM sodium chloride, 100mM imidazole, 0.5mM PMSF, 1mM DTT and 10% glycerol. |