missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Suppressor of cytokine signaling 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Suppressor of cytokine signaling 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6712-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001257396.1 | |
Suppressor of cytokine signaling 2 Protein | |
Research Use Only | |
SOCS2 | |
Recombinant Protein | |
E. coli | |
MTLRCLEPSG NGGEGTRSQW GTAGSAEEPS PQAARLAKAL RELGQTGWYW GSMTVNEAKE KLKEAPEGTF LIRDSSHSDY LLTISVKTSA GPTNLRIEYQ DGKFRLDSII CVKSKLKQFD SVVHLIDYYV QMCKDKRTGP EAPRNGTVHL YLTKPLYTSA PSLQHLCRLT INKCTGAIWG LPLPTRLKDY LEEYKFQV | |
Greater than 80% as determined by SDS-PAGE. |
8835 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |