missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Suppressor of cytokine signaling 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Suppressor of cytokine signaling 2, was expressed in the hosts and tags indicated.
245.00 € - 3260.00 €
Specifications
Accession Number | NP_001257396.1 |
---|---|
Gene ID (Entrez) | 8835 |
Name | Suppressor of cytokine signaling 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16035300
|
enQuireBio™
QP6712-EC-100UG |
100 μg |
735.00 €
100µg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16055300
|
enQuireBio™
QP6712-EC-1MG |
1 mg |
3260.00 €
1mg |
Estimated Shipment: 21-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16045300
|
enQuireBio™
QP6712-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065300
|
enQuireBio™
QP6712-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16075300
|
enQuireBio™
QP6712-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085300
|
enQuireBio™
QP6712-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001257396.1 | |
Suppressor of cytokine signaling 2 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
8835 | |
Research Use Only | |
SOCS2 | |
Recombinant Protein | |
E. coli | |
MTLRCLEPSG NGGEGTRSQW GTAGSAEEPS PQAARLAKAL RELGQTGWYW GSMTVNEAKE KLKEAPEGTF LIRDSSHSDY LLTISVKTSA GPTNLRIEYQ DGKFRLDSII CVKSKLKQFD SVVHLIDYYV QMCKDKRTGP EAPRNGTVHL YLTKPLYTSA PSLQHLCRLT INKCTGAIWG LPLPTRLKDY LEEYKFQV | |
Greater than 80% as determined by SDS-PAGE. |