missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Survivin / BIRC5 / API4 Protein
A DNA sequence encoding the Homo sapiens (Human) Survivin / BIRC5 / API4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5719-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_001012270.1 | |
Survivin / BIRC5 / API4 Protein | |
Research Use Only | |
BIRC5 | |
Recombinant Protein | |
E. coli | |
MGAPTLPPAW QPFLkDaHRI STFKNWPFLE GCACTPERMA EAGFIHCPTE NEPDLAQCFF CFKELEGWEP DDDPIEEHKK HSSGCAFLSV KKQFEELTLG EFLKLDRERA KNKIAKETNN KKKEFEETAK KVRRAIEQLA AMD | |
Greater than 80% as determined by SDS-PAGE. |
332 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |