missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SVIP Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
10 μg
2 μg
Förpackningsstorlek
10µg
1mg
2µg
Tekniske data
Tekniske data
| Name | SVIP Protein |
| Quantity | 1 mg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MGSMGLCFPC PGESAPPTPD LEEKRAKLAE AAERRQKEAA SRGILDVQSV QEKRKKKEKI EKQIATSGPP PEGGLRWTVS |
| Buffer | The SVIP solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 2mM DTT. |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion