missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human SYK Protein

A cDNA sequence encoding the SYK was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13656-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934289

  • 1450.00 € / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

6850
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Human (HEK293)
MHHHHHHDYKDDDDKKLMASSGMADSANHLPFFFGNITREEAEDYLVQGGMSDGL YLLRQSRNYLGGFALSVAHGRKAHHYTIERELNGTYAIAGGRTHASPADLCHYHSQES DGLVCLLKKPFNRPQGVQPKTGPFEDLKENLIREYVKQTWNLQGQALEQAIISQKPQL EKLIATTAHEKMPWFHGKISREESEQIVLIGSKTNGKFLIRARDNNGSYALCLLHEGKV LHYRIDKDKTGKLSIPEGKKFDTLWQLVEHYSYKADGLLRVLTVPCQKIGTQGNVNFG GRPQLPGSHPATWSAGGIISRIKSYSFPKPGHRKSSPAQGNRQESTVSFNPYEPELA PWAADKGPQREALPMDTEVYESPYADPEEIRPKEVYLDRKLLTLEDKELGSGNFGTV KKGYYQMKKVVKTVAVKILKNEANDPALKDELLAEANVMQQLDNPYIVRMIGICEAES WMLVMEMAELGPLNKYLQQNRHVKDKNIIELVHQVSMGMKYLEESNFVHRDLAARN VLLVTQHYAKISDFGLSKALRADENYYKAQTHGKWPVKWYAPECINYYKFSSKSDVW SFGVLMWEAFSYGQKPYRGMKGSEVTAMLEKGERMGCPAGCPREMYDLMNLCWT YDVENRPGFAAVELRLRNYYYDVVN
Greater than 85.0% as determined by SDS-PAGE.
SYK Protein
Research Use Only
SYK
Human
Flag Tag
SYK protein is supplied in 50mM Tris pH 7.5, 300mM NaCl and 10% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers