missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TAF12 Protein

A DNA sequence encoding the Homo sapiens (Human) TAF12, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7622-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16039850

  • 505.00 € / 100µg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001128690.1
TAF12 Protein
Research Use Only
TAF12
Recombinant Protein
E. coli
MNQFGPSALI NLSNFSSIKP EPASTPPQGS MANSTAVVKI PGTPGAGGRL SPENNQVLTK KKLQDLVREV DPNEQLDEDV EEMLLQIADD FIESVVTAAC QLARHRKSST LEVKDVQLHL ERQWNMWIPG FGSEEIRPYK KACTTEAHKQ RMALIRKTTK K
Greater than 90% as determined by SDS-PAGE.
6883
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers