missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TAF15 Protein
A cDNA sequence encoding the TAF15 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13685-20ug
Additional Details : Weight : 0.01000kg
Specifications
TAF15 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSSYHSQRE NYSHHTQDDR RDVSRYGEDN RGYGGSQGGG RGRGGYDKDG RGPMTGSSGG DRGGFKNFGG HRDYGPRTDA DSESDNSDNN TIFVQGLGEG VSTDQVGEFF KQIGIIKTNK KTGKPMINLY TDKDTGKPKG EATVSFDDPP SAKAAIDWFD GKEFHGNIIK VSFATRRPEF MRGGGSGGGR RGRGGYRGRG GFQGRGGDPK SGDWVCPNPS CGNMNFARRN SCNQCNEPRP EDSRPSGGDF RGRGYGGERG YR | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TAF15 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT. |