missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TAF15 Protein

A cDNA sequence encoding the TAF15 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13685-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954379

  • 131.00 € / 5µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TAF15 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSSYHSQRE NYSHHTQDDR RDVSRYGEDN RGYGGSQGGG RGRGGYDKDG RGPMTGSSGG DRGGFKNFGG HRDYGPRTDA DSESDNSDNN TIFVQGLGEG VSTDQVGEFF KQIGIIKTNK KTGKPMINLY TDKDTGKPKG EATVSFDDPP SAKAAIDWFD GKEFHGNIIK VSFATRRPEF MRGGGSGGGR RGRGGYRGRG GFQGRGGDPK SGDWVCPNPS CGNMNFARRN SCNQCNEPRP EDSRPSGGDF RGRGYGGERG YR
Greater than 90% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TAF15 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers