missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TCEAL7 Protein

A cDNA sequence encoding the TCEAL7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13708-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934449

  • 203.00 € / 20µg
Estimated Shipment: 24-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TCEAL7 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMQKPCK ENEGKPKCSV PKREEKRPYG EFERQQTEGN FRQRLLQSLE EFKEDIDYRH FKDEEMTREG DEMERCLEEI RGLRKKFRAL HSNHRHSRDR PYPI
Greater than 80% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The TCEAL7 solution (0.5 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 50% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers