missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TCF4 Protein
A cDNA sequence encoding the TCF4 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13710-HIS-INSECT-2ug
Additional Details : Weight : 0.01000kg
Specifications
TCF4 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
MGSSHHHHHH SSGLVPRGSH MGSMKFKQCR CSDTGLCCLD HEGKAEVYAP SASTADYNRD SPGYPSSKPA TSTFPSSFFM QDGHHSSDPW SSSSGMNQPG YAGMLGNSSH IPQSSSYCSL HPHERLSYPS HSSADINSSL PPMSTFHRSG TNHYSTSSCT PPANGTDSIM ANRGSGAAGS SQTGDALGKA LASIYSPDHT NNSFSSNPST PVGSPPSLSA GTAVWSRNGG QASSSPNYEG PLHSLQSRIE DRLERLDDAI HVLRNHAVGP STAMPGGHGD MHGIIGPSHN GAMGGLGSGY GTGLLSANRH SLMVGTHRED GVALRGSHSL LPNQVPVPQL PVQSATSPDL NPPQDPYRGM PPGLQGQSVS SGSSEIKSDD EGDENLQDTK SSEDKKLDDD KKDIKSITSN NDDEDLTPEQ KAEREKERRM ANNARERLRV RDINEAFKEL GRMVQLHLKS DKPQTKLLIL HQAVAVILSL EQQVRERNLN PKAACLKRRE EEKVSSEPPP LSLAGPHPGM GDASNHMGQM | |
Greater than 90.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The TCF4 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.1M NaCl. |