missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TCL1B Protein
A cDNA sequence encoding the TCL1B was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13712-20ug
Additional Details : Weight : 0.01000kg
Specifications
TCL1B Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASEASV RLGVPPGRLW IQRPGIYEDE EGRTWVTVVV RFNPSRREWA RASQGSRYEP SITVHLWQMA VHTRELLSSG QMPFSQLPAV WQLYPGRKYR AADSSFWEIA DHGQIDSMEQ LVLTYQPERK D | |
Greater than 95% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TCL1B protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl, 40% glycerol and 2mM DTT. |