missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human TGF-beta 1 Biotinylated Protein

Product Code. 18757106
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18757106 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18757106 Supplier R&D Systems™ Supplier No. BT7754025
  • ONLINE EXCLUSIVE PRICE
    1284.00 €
    1144.50 € / 25µg
    Save 139.50 €
    10% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human Cell-expressed, Biotinylated

Measured by its binding ability in a functional ELISA. When Recombinant Human TGF-beta RII Fc Chimera (Catalog # 341-BR ) is immobilized at 0.1 μg/mL (100 μL/well), Biotinylated Recombinant Human TGF-beta 1 (Catalog # BT7754) binds with an ED50 of 1.50-15.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01137.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Acetonitrile and TFA with Trehalose.
Gene ID (Entrez) 7040
Molecular Weight (g/mol) M.W.-observed: 8-14 kDa, under reducing conditions., M.W.-theroretical: 12.8 kDa
Quantity 25 μg
Source Human embryonic kidney cell, HEK293-derived human TGF-beta 1 protein Ala279-Ser390
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CEDLAP, DPD1, latency-associated peptide, TGF beta, TGF beta1, TGFB, TGFB1, TGFbeta, TGFbeta 1, TGF-beta 1 protein, TGF-beta-1, transforming growth factor beta-1, transforming growth factor, beta 1
Conjugate Biotin
Reconstitution Reconstitute at 250 μg/mL in 4 mM HCl.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein TGF-beta 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.