missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TGF-beta 3, Animal-Free Protein
Click to view available options
Quantity:
50 μg
500 μg
Unit Size:
500µg
50µg
Description
No significant difference between EC50 of reference and test lots
Specifications
Specifications
| Accession Number | P10600.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from acetonitrile/TFA |
| Gene ID (Entrez) | 7043 |
| Molecular Weight (g/mol) | M.W. (Observed): Dimeric TGF-beta 3 protein only, M.W. (Theroretical): 12.7 kDa (monomer), 25.4 kDa (dimer) |
| Quantity | 50 μg |
| Source | E. coli-derived human TGF-beta 3 protein |
| Storage Requirements | Store lyophilized protein between -20°C to -80°C until the date of expiry. Avoid freeze-thaw cycles. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | ARVD, FLJ16571, TGF-beta3, TGF-beta-3, transforming growth factor beta-3, transforming growth factor, beta 3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction