missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human TGF-beta 3 (CHO-expressed) Protein
Click to view available options
Quantity:
25 μg
5 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein
Unit Size:
25µg
5µg
Description
The Recombinant Human TGF-beta 3 (CHO-expressed) Protein is derived from CHO. The Recombinant Human TGF-beta 3 (CHO-expressed) Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | P10600 |
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA |
| Gene ID (Entrez) | 7043 |
| Molecular Weight (g/mol) | 13kDa |
| Name | TGF-beta 3 |
| Preparation Method | Reconstitute at 50μg/mL in 4mM HCl. |
| Quality Control Testing | SDS-PAGE: 9-11kDa, reducing conditions |
| Quantity | 25 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived human TGF-beta 3 protein Ala301-Ser412 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction