missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TGFB3 Protein

Recombinant Protein

Brand:  enQuireBio™ QP5237-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15965759

  • 252.00 € / 10µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001316867.1
TGFB3 Protein
Research Use Only
TGFB3
Human
Untagged
10 mM acetic acid and 20% Ethanol at a concentration of 0.25 mg/mL
7043
10 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS
Greater than 95% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers