missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGFB3 Protein
Recombinant Protein
Brand: enQuireBio™ QP5237-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001316867.1 | |
TGFB3 Protein | |
Research Use Only | |
TGFB3 | |
Human | |
Untagged | |
10 mM acetic acid and 20% Ethanol at a concentration of 0.25 mg/mL |
7043 | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MALDTNYCFR NLEENCCVRP LYIDFRQDLG WKWVHEPKGY YANFCSGPCP YLRSADTTHS TVLGLYNTLN PEASASPCCV PQDLEPLTIL YYVGRTPKVE QLSNMVVKSC KCS | |
Greater than 95% as determined by SDS-PAGE. |