missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TGFBI (135 Amino Acids Protein

A cDNA sequence encoding the TGFBI (135 Amino Acids was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13733-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934529

  • 203.00 € / 20µg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TGFBI (135 Amino Acids Protein
Research Use Only
Recombinant Protein
E. coli
MGTVMDVLKG DNRFSMLVAA IQSAGLTETL NREGVYTVFA PTNEAFRALP PRERSRLLGD AKELANILKY HIGDEILVSG GIGALVRLKS LQGDKLEVSL KNNVVSVNKE PVAEPDIMAT NGVVHVITNV LQPPA
Greater than 95.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
The TGFBI recombinant Human is formulated 20mM Tris-HCl pH 8.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers