missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Thrombopoietin (E. coli-expressed)

Product Code. 18768876
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18768876 50 μg 50µg
18717416 10 μg 10µg
18706146 1 mg 1mg
18726986 100 μg 100µg
18729306 250 μg 250µg
18707726 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 18768876 Supplier R&D Systems™ Supplier No. 288TPE050
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A key regulator of megakaryocytopoiesis and thrombopoiesis

The Recombinant Human Thrombopoietin (E. coli-expressed) is derived from E. coli. The Recombinant Human Thrombopoietin (E. coli-expressed) has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_000451
For Use With (Application) Bioactivity
Gene ID (Entrez) 7066
Molecular Weight (g/mol) 18.7 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source E. coli-derived human Thrombopoietin/Tpo protein Ser22-Leu195
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, ≤ -20°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 100 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Product Line R&D Systems
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.