missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Thrombopoietin His (HEK-293) Protein

Code produit 18765775
Modifier l'affichage
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
Conditionnement:
100µg
10µg
1mg
500µg
50µg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Conditionnement
18765775 50 μg 50µg
18745725 500 μg 500µg
18706175 100 μg 100µg
18786175 1 mg 1mg
18735885 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18765775 Fournisseur R&D Systems™ Code fournisseur 288TPH050
 Restricted Item
Estimated Shipment: 02-12-2026
Ajouter au panier
Ajouter au panier

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Analyzed by SEC-MALS. His-tag (HEK-293-expressed)

Measured in a cell proliferation assay using MO7e human megakaryocytic leukemic cells. Avanzi, G. et al. (1988) Br. J. Haematol. 69:359. The ED50 for this effect is 0.0500-0.500 ng/mL.
TRUSTED_SUSTAINABILITY

Spécification

Accession Number P40225.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 7066
Molecular Weight (g/mol) MolecularWeight-observed: 26-30 kDa, under reducing conditions., MolecularWeight-theroretical: 19 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Thrombopoietin/Tpo protein Ser22-Leu195, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias Megakaryocyte colony-stimulating factor, Megakaryocyte growth and development factor, megakaryocyte stimulating factor, MGDF, MGDFC-mpl ligand, MKCSF, MK-CSF, ML, MPL ligand, MPLLG, MPLLGMGC163194, Myeloproliferative leukemia virus oncogene ligand, THCYT1, THPO, thrombopoietin, thrombopoietin nirs variant 1, Tpo, TPOMKCSF
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes (at 500 μg/mL in PBS.)
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Thrombopoietin/THPO
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.