missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Thyroid peroxidase / TPO Protein

A DNA sequence encoding the Homo sapiens (Human) Thyroid peroxidase / TPO, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9441-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038321

  • 1665.00 € / 500µg
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000538.3
Thyroid peroxidase / TPO Protein
Research Use Only
TPO
Recombinant Protein
Yeast
FFPFISRGKE LLWGKPEESR VSSVLEESKR LVDTAMYATM QRNLKKRGIL SPAQLLSFSK LPEPTSGVIA RAAEIMETSI QAMKRKVNLK TQQSQHPTDA LSEDLLSIIA NMSGCLPYML PPKCPNTCLA NKYRPITGAC NNR
Greater than 90% as determined by SDS-PAGE.
7173
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers