missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TIMM8A Protein
A DNA sequence encoding the Homo sapiens (Human) TIMM8A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6794-ec-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004076.1 | |
TIMM8A Protein | |
Research Use Only | |
TIMM8A | |
Recombinant Protein | |
E. coli | |
MDSSSSSSAA GLGAVDPQLQ HFIEVETQKQ RFQQLVHQMT ELCWEKCMDK PGPKLDSRAE ACFVNCVERF IDTSQFILNR LEQTQKSKPV FSESLSD | |
Greater than 80% as determined by SDS-PAGE. |
1678 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |