missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TK2 Protein

A cDNA sequence encoding the TK2 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13757-2ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934609

  • 131.00 € / 2µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TK2 Protein
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMVQRRA WPPDKEQEKE KKSVICVEGN IASGKTTCLE FFSNATDVEV LTEPVSKWRN VRGHNPLGLM YHDASRWGLT LQTYVQLTML DRHTRPQVSS VRLMERSIHS ARYIFVENLY RSGKMPEVDY VVLSEWFDWI LRNMDVSVDL IVYLRTNPET CYQRLKKRCR EEEKVIPLEY LEAIHHLHEE WLIKGSLFPM AAPVLVIEAD HHMERMLELF EQNRDRILTP ENRKHCP
Greater than 85% as determined by SDS-PAGE.
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
TK2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0, 30% glycerol, 2mM DTT and 200mM NaCl.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers