missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNFAIP8 Protein

A DNA sequence encoding the Homo sapiens (Human) TNFAIP8, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8735-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16046221

  • 2285.00 € / 1mg
Estimated Shipment: 24-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001071122.1
TNFAIP8 Protein
Research Use Only
TNFAIP8
Recombinant Protein
E. coli
HSEAEESKEV ATDVFNSKNL AVQAQKKILG KMVSKSIATT LIDDTSSEVL DELYRVTREY TQNKKEAEKI IKNLIKTVIK LAILYRNNQF NQDELALMEK FKKKVHQLAM TVVSFHQVDY TFDRNVLSRL LNECREMLHQ IIQRHLTAKS HGRVNNVFDH FSDCEFLAAL YNPFGNFKPH LQKLCDGINK MLDEENI
Greater than 90% as determined by SDS-PAGE.
25816
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers