missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNFRSF10A Protein

A cDNA sequence encoding the TNFRSF10A was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10887-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15905478

  • 203.00 € / 50µg
Estimated Shipment: 21-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

8797
50 μg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50, as measured by its ability to inhibit Trail-mediated cytotoxicity in mouse L-929 cells, is less than 1.25ng/ml.
Human
Fc Tag
Lyophilized from a 0.2microm filtered solution in PBS.
TNFRSF10A Protein
Research Use Only
TNFRSF10A
Recombinant Protein
E. coli
ASGTEAAAAT PSKVWGSSAG RIEPRGGGRG ALPTSMGQHG PSARARAGRA PGPRPAREAS PRLRVHKTFK FVVVGVLLQV VPSSAATIKL HDQSIGTQQW EHSPLGELCP PGSHRSERPG ACNRCTEGVG YTNASNNLFA CLPCTACKSD EEERSPCTTT RNTACQCKPG TFRNDNSAEM CRKCSTGCPR GMVKVKDCTP WSDIECVHKE SGNGHNIEGR MDPKSCDKTH TCPPCPAPEL LGGPSVFLFP PKPKDTLMIS RTPEVTCVVV DVSHEDPEVK FNWYVDGVEV HNAKTKPREE QYNSTYRVVS VLTVLHQDWL NGKEYKCKVS NKALPAPIEK TISKAKGQPR EPQVYTLPPS RDELTKNQVS LTCLVKGFYP SDIAVEWESN GQPENNYKTT PPVLDSDGSF FLYSKLTVDK SRWQQGNVFS CSVMHEALHN HYTQKSLSLS PGK
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers