missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TNFRSF8 Protein

A cDNA sequence encoding the TNFRSF8 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13771-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934649

  • 203.00 € / 10µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

943
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
Baculovirus
ADPFPQDRPF EDTCHGNPSH YYDKAVRRCC YRCPMGLFPT QQCPQRPTDC RKQCEPDYYL DEADRCTACV TCSRDDLVEK TPCAWNSSRV CECRPGMFCS TSAVNSCARC FFHSVCPAGM IVKFPGTAQK NTVCEPASPG VSPACASPEN CKEPSSGTIP QAKPTPVSPA TSSASTMPVR GGTRLAQEAA SKLTRAPDSP SSVGRPSSDP GLSPTQPCPE GSGDCRKQCE PDYYLDEAGR CTACVSCSRD DLVEKTPCAW NSSRTCECRP GMICATSATN SCARCVPYPI CAAETVTKPQ DMAEKDTTFE APPLGTQPDC NPTPENGEAP ASTSPTQSLL VDSQASKTLP IPTSAPVALS STGKHHHHHH
Greater than 95.0% as determined by SDS-PAGE.
TNFRSF8 Protein
Research Use Only
TNFRSF8
Human
His
TNFRSF8 protein solution (1 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers