missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TnT Protein

A cDNA sequence encoding the TnT was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13779-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15954679

  • 131.00 € / 10µg
Estimated Shipment: 26-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

TnT Protein
Research Use Only
Recombinant Protein
Human
MSDIEEVVEE YEEEEQEEAA VEEEEDWRED EDEQEEAAEE DAEAEAETEE TRAEEDEEEE EAKEAEDGPM EESKPKPRSF MPNLVPPKIP DGERVDFDDI HRKRMEKDLN ELQALIEAHF ENRKKEEEEL VSLKDRIERR RAERAEQQRI RNEREKERQN RLAEERARRE EEENRRKAED EARKKKALSN MMHFGGYIQK QAQTERKSGK RQTEREKKKK ILAERRKVLA IDHLNEDQLR EKAKELWQSI YNLEAEKFDL QEKFKQQKYE INVLRNRIND NQKVSKTRGK AKVTGRWK
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
Human Cardiac Troponin T in 6M Urea, 250mM NaCl, 50mM Tris-HCl, 15mM b-mercaptoethanol, 1mM EDTA, pH 7.2.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers