missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TnT Protein
A cDNA sequence encoding the TnT was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13779-50ug
Additional Details : Weight : 0.01000kg
Specifications
TnT Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
MSDIEEVVEE YEEEEQEEAA VEEEEDWRED EDEQEEAAEE DAEAEAETEE TRAEEDEEEE EAKEAEDGPM EESKPKPRSF MPNLVPPKIP DGERVDFDDI HRKRMEKDLN ELQALIEAHF ENRKKEEEEL VSLKDRIERR RAERAEQQRI RNEREKERQN RLAEERARRE EEENRRKAED EARKKKALSN MMHFGGYIQK QAQTERKSGK RQTEREKKKK ILAERRKVLA IDHLNEDQLR EKAKELWQSI YNLEAEKFDL QEKFKQQKYE INVLRNRIND NQKVSKTRGK AKVTGRWK | |
Greater than 95.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Human Cardiac Troponin T in 6M Urea, 250mM NaCl, 50mM Tris-HCl, 15mM b-mercaptoethanol, 1mM EDTA, pH 7.2. |