missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TOM1L2 Protein
A cDNA sequence encoding the TOM1L2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13781-10ug
Additional Details : Weight : 0.01000kg
Specifications
TOM1L2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSEFMEFLL GNPFSTPVGQ CLEKATDGSL QSEDWTLNME ICDIINETEE GPKDAIRALK KRLNGNRNYR EVMLALTVLE TCVKNCGHRF HILVANRDFI DSVLVKIISP KNNPPTIVQD KVLALIQAWA DAFRSSPDLT GVVHIYEELK RKGVEFPMADLDALSPIHTP QRSVPEVDPA ATMPRSQSQQ RTSAGSYSSP PPAPYSAPQA PALSVTGPIT ANSEQIARLR SELDVVRGNT KVMSEMLTEM VPGQEDSSDL ELLQELNRTC RAMQQRIVEL ISRVSNEEVT EELLHVNDDL NNVFLRYERF ERYRSGRSVQ NASNGVLNEV TEDNLIDLGP GSPAVVSPMV GNTAPPSSLS SQLAGLDLGT ESVSGTLSSL QQCNPRDGFD MFAQTRGNSL AEQRKTVTYE DPQAVGGLAS ALDNRKQSSE GIPVAQPSVM DDIEVWLRTD LKGDDLEEGV TSEEFDKFLE ERAKAAEMVP DLPSPPMEAP APASNPSGRK KPERSEDALF AL | |
Greater than 80.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
TOM1L2 protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol and 1mM DTT. |