missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TOP1 Protein
A cDNA sequence encoding the TOP1 was constructed and used to recombinantly synthesize the protein.
131.00 € - 5655.00 €
Specifications
Gene ID (Entrez) | 7150 |
---|---|
Name | TOP1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Gene Symbol | TOP1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15994689
|
enQuireBio™
QP13783-5UG |
5 μg |
131.00 €
5µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15984689
|
enQuireBio™
QP13783-20UG |
20 μg |
203.00 €
20µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15974689
|
enQuireBio™
QP13783-1MG |
1 mg |
5655.00 €
1mg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
7150 | |
Research Use Only | |
TOP1 | |
Human | |
His | |
TOP1 is supplied in 16mM HEPES buffer pH 7.5, 400mM sodium chloride, and 20% glycerol. |
TOP1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
MSGDHLHNDS QIEADFRLND SHKHKDKHKD REHRHKEHKK EKDREKSKHS NSEHKDSEKK HKEKEKTKHK DGSSEKHKDK HKDRDKEKRK EEKVRASGDA KIKKEKENGF SSPPQIKDEP EDDGYFVPPK EDIKPLKRPR DEDDADYKPK KIKTEDTKKE KKRKLEEEED GKLKKPKNKD KDKKVPEPDN KKKKPKKEEE QKWKWWEEER YPEGIKWKFL EHKGPVFAPP YEPLPENVKF YYDGKVMKLS PKAEEVATFF AKMLDHEYTT KEIFRKNFFK DWRKEMTNEE KNIITNLSKC DFTQMSQYFK AQTEARKQMS KEEKLKIKEE NEKLLKEYGF CIMDNHKERI ANFKIEPPGL FRGRGNHPKM GMLKRRIMPE DIIINCSKDA KVPSPPPGHK WKEVRHDNKV TWLVSWTENI QGSIKYIMLN PSSRIKGEKD WQKYETARRL KKCVDKIRNQ YREDWKSKEM KVRQRAVALY FIDKLALRAG NEKEEGETAD TVGCCSLRVE HINLHPELDG QEYVVEFDFL GKDSIRYYNK VPVEKRVFKN LQLFMENKQP EDDLFDRLNT GILNKHLQDL MEGLTAKVFR TYNASITLQQ QLKELTAPDE NIPAKILSYN RANRAVAILC NHQRAPPKTF EKSMMNLQTK IDAKKEQLAD ARRDLKSAKA DAKVMKDAKT KKVVESKKKA VQRLEEQLMK LEVQATDREE NKQIALGTSK LNYLDPRITV AWCKKWGVPI EKIYNKTQRE KFAWAIDMAD EDYEF | |
Greater than 95% as determined by SDS-PAGE. |