missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Transcriptional coactivator YAP1 Protein

A DNA sequence encoding the Homo sapiens (Human) Transcriptional coactivator YAP1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9470-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16038381

  • 1730.00 € / 500µg
Estimated Shipment: 05-08-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001123617.1
Transcriptional coactivator YAP1 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
10413
Greater than 90% as determined by SDS-PAGE.
Research Use Only
YAP1
Recombinant Protein
Yeast
MDPGQQPPPQ PAPQGQGQPP SQPPQGQGPP SGPGQPAPAA TQAAPQAPPA GHQIVHVRGD SETDLEALFN AVMNPKTANV PQTVPMRLRK LPDSFFKPPE PKSHSRQAST DAGTAGALTP QHVRAHSSPA SLQLGAVSPG TLTPTGVVSG PAATPTAQHL RQSSFEIPDD VPLPAGWEMA KTSSGQRYFL NHIDQTTTWQ DPRKAMLSQM NVTAPTSPPV QQNMMNSASG PLPDGWEQAM TQDGEIYYIN HKNKTTSWLD PRLDPRFAMN QRISQSAPVK QPPPLAPQSP QGGVMGGSNS NQQQQMRLQQ LQMEKERLRL KQQELLRQAM RNINPSTANS PKCQELALRS QLPTLEQDGG TQNPVSSPGM SQELRTMTTN SSDPFLNSGT YHSRDESTDS GLSMSSYSVP RTPDDFLNSV DEMDTGDTIN QSTLPSQQNR FPDYLEAIPG TNVDLGTLEG DGMNIEGEEL MPSLQEALSS DILNDMESVL AATKLDKESF LTWL
Videos
SDS
Documents
Certificates
Special Offers

Special Offers