missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transcriptional enhancer factor TEF-1 Protein
A DNA sequence encoding the Homo sapiens (Human) Transcriptional enhancer factor TEF-1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9426-ye-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_068780.2 | |
Transcriptional enhancer factor TEF-1 Protein | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7003 | |
Greater than 90% as determined by SDS-PAGE. | |
Research Use Only | |
TEAD1 | |
Recombinant Protein | |
Yeast | |
MEPSSWSGSE SPAENMERMS DSADKPIDND AEGVWSPDIE QSFQEALAIY PPCGRRKIIL SDEGKMYGRN ELIARYIKLR TGKTRTRKQV SSHIQVLARR KSRDFHSKLK DQTAKDKALQ HMAAMSSAQI VSATAIHNKL GLPGIPRPTF PGAPGFWPGM IQTGQPGSSQ DVKPFVQQAY PIQPAVTAPI PGFEPASAPA PSVPAWQGRS IGTTKLRLVE FSAFLEQQRD PDSYNKHLFV HIGHANHSYS DPLLESVDIR QIYDKFPEKK GGLKELFGKG PQNAFFLVKF WADLNCNIQD DAGAFYGVTS QYESSENMTV TCSTKVCSFG KQVVEKVETE YARFENGRFV YRINRSPMCE YMINFIHKLK HLPEKYMMNS VLENFTILLV VTNRDTQETL LCMACVFEVS NSEHGAQHHI YRLVKD |